missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Sulfate, Anhydrous, Powder, BAKER ANALYZED™ Reagent, J.T. Baker™

Product Code. 15146376 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15146376 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15146376 Supplier Avantor J.T.Baker Supplier No. 250601
  • ONLINE EXCLUSIVE PRICE
    225.78 €
    173.84 € / 500g
    Save 51.94 €
    23% Off
 Restricted Item
Estimated Shipment: 08-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 7487-88-9
Physical Form Powder
Quantity 500 g
Packaging Poly Bottle
Specific Gravity 1L = 2.65kg
Molecular Formula MgO4S
MDL Number MFCD00011110
Synonym magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
SMILES [Mg++].[O-]S([O-])(=O)=O
IUPAC Name magnesium;sulfate
Molecular Weight (g/mol) 120.36
PubChem CID 24083
ChEBI CHEBI:32599
Formula Weight 120.37
Percent Purity ≥99.0%
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Magnesium Sulfate, Anhydrous
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.