missing translation for 'onlineSavingsMsg'
Learn More

Magnesium sulfate, dried, contains ca 1-2 mol water of hydration

Product Code. 11393207
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11393207 2500 g 2.5kg
11373207 100 g 100g
11383207 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11393207 Supplier Thermo Scientific Alfa Aesar Supplier No. L13739.0E
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 22189-08-8 | MgO4S | 120.36 g/mol

Suitable for the preliminary drying of organic liquids, for example solvent extracts from reaction workups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22189-08-8
Molecular Formula MgO4S
Molecular Weight (g/mol) 120.36
MDL Number MFCD00149787
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
Synonym magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
PubChem CID 24083
ChEBI CHEBI:32599
IUPAC Name magnesium;sulfate
SMILES [Mg++].[O-]S([O-])(=O)=O

Specifications

Quantity 2500 g
Merck Index 14,5691
Formula Weight 120.37 (Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Magnesium sulfate, contains ≈1 to 2 mol water of hydration

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.