missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Sulfate, Extra Pure, SLR, Dried, Fisher Chemical™

Product Code. 10181860 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2 kg
10 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
10kg
1kg
2kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10181860 10 kg Plastic keg 10kg
10316240 500 g Plastic powder jar 500g
10776101 1 kg Plastic powder jar 1kg
10224680 2 kg Plastic powder jar 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10181860 Supplier Fisher Chemical Supplier No. M/1100/68
Spotlight Offers
Use promo code 28036
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
This item is not returnable. View return policy

Magnesium Sulfate, Extra Pure, SLR, Dried

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22189-08-8
Molecular Formula MgO4S
Molecular Weight (g/mol) 120.36
MDL Number MFCD00149787
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
Synonym magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
PubChem CID 24083
ChEBI CHEBI:32599
SMILES [Mg++].[O-]S([O-])(=O)=O

Specifications

Melting Point 1124°C
Color White
Physical Form Powder/Solid
Quantity 10 kg
IUPAC Name magnesium(2+) sulfate
Formula Weight 120.36g/mol
Packaging Plastic keg
Vapor Pressure <0.1mmHg at 20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.