missing translation for 'onlineSavingsMsg'
Learn More

Magnesium sulfate heptahydrate, 99.5%, for analysis

Product Code. 10378703 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
Unit Size:
2.5kg
25kg
500g
Product Code. Quantity Packaging unitSize
10378703 25 kg Fibre drum 25kg
10256840 500 g Plastic bottle 500g
10135453 2.5 kg Plastic bottle 2.5kg
3 options
This item is not returnable. View return policy

Product Code. 10378703

Brand: Thermo Scientific Acros 213110250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10034-99-8 | H14MgO11S | 246.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-99-8
Molecular Formula H14MgO11S
Molecular Weight (g/mol) 246.47
MDL Number MFCD00149785
InChI Key WRUGWIBCXHJTDG-UHFFFAOYSA-L
Synonym magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
PubChem CID 24843
ChEBI CHEBI:31795
IUPAC Name magnesium;sulfate;heptahydrate
SMILES O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

Specifications

Melting Point 770.0°C
Color White
pH 5 to 8 (5% soln. at 25°C)
Physical Form Crystals
Quantity 25 kg
Assay Percent Range 99.4% min. (Complexometry)
Linear Formula MgSO4·7H2O
Merck Index 15, 5755
Solubility Information Solubility in water: 710g/L (20°C). Other solubilities: soluble in glycerol,sparingly soluble in alcohol
Formula Weight 246.48
Percent Purity 99.5%
Grade Analytical
Packaging Fibre drum
Chemical Name or Material Magnesium sulfate heptahydrate, p.a.
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.