missing translation for 'onlineSavingsMsg'
Learn More

Magnesium sulfate heptahydrate, 99+%

Product Code. p-7021946
Click to view available options
Quantity:
1000 g
5000 g
Unit Size:
1000g
5000g
This item is not returnable. View return policy

Product Code. 11411828

Brand: Thermo Scientific Alfa Aesar A14491.0B

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10034-99-8 | H14MgO11S | 246.47 g/mol

Magnesium sulfate, produced by appropriate processes, finds use in several pharmaceutical preparations of magnesium usually known as epsom salt. It is used as a micronutrient in agriculture, for brewing salt in the production of certain alcoholic products, as an electrolyte to prepare metal salts, as a coagulant and as an ingredient in bath salts for use in floatation therapy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium sulfate, produced by appropriate processes, finds use in several pharmaceutical preparations of magnesium usually known as epsom salt. It is used as a micronutrient in agriculture, for brewing salt in the production of certain alcoholic products, as an electrolyte to prepare metal salts, as a coagulant and as an ingredient in bath salts for use in floatation therapy.

Solubility
Soluble in in water and acid. Slightly soluble in alcohol and glycerol.

Notes
Incompatible with strong oxidizers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-99-8
Molecular Formula H14MgO11S
Molecular Weight (g/mol) 246.47
MDL Number MFCD00149785
InChI Key WRUGWIBCXHJTDG-UHFFFAOYSA-L
Synonym magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
PubChem CID 24843
ChEBI CHEBI:31795
IUPAC Name magnesium;sulfate;heptahydrate
SMILES O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

Specifications

Quantity 1000 g
Merck Index 14,5691
Solubility Information Soluble in in water and acid. Slightly soluble in alcohol and glycerol.
Formula Weight 246.48 (120.37 Anhydrous)
Percent Purity ≥99%
Odor Odorless
Density 1.68
Chemical Name or Material Magnesium sulfate heptahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.