missing translation for 'onlineSavingsMsg'
Learn More

Magnesium sulfate heptahydrate, ACS Reagent, ≥99.0%, Solstice

Product Code. 16695873
Change view
Click to view available options
Quantity:
5 kg
Unit Size:
5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16695873 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16695873 Supplier Fluka Supplier No. 631405KG
  • ONLINE EXCLUSIVE PRICE
    350.00 €
    316.15 € / 5kg
    Save 33.85 €
    9% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Boiling Point Not Determined
Melting Point 1,124°C (Decomposes before melting)
CAS 10034-99-8
Color Colorless
CAS Max % 1
pH 5 to 9.2
Quantity 5 kg
Molecular Formula H14MgO11S
InChI Key WRUGWIBCXHJTDG-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O
IUPAC Name magnesium(2+) heptahydrate sulfate
Molecular Weight (g/mol) 246.47
Density 1.68 g/cm3 (20°C)
Chemical Name or Material Magnesium sulfate heptahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.