missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Sulfate Heptahydrate, Certified AR for Analysis, Fisher Chemical™

Product Code. 10346190 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10346190 1 kg 1kg
10697412 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10346190 Supplier Fisher Chemical Supplier No. M/1050/60
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Magnesium Sulfate Heptahydrate, Certified AR for Analysis, Fisher Chemical, Quantity: 500g, CAS: 10034-99-8, White, Melting Point: 770 deg.C, Molecular Formula: H14MgO11S, Molecular Weight: 246.466, Packaging: Plastic powder jar, pH: 5 to 8.2

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-99-8
Molecular Formula H14MgO11S
Molecular Weight (g/mol) 246.47
MDL Number MFCD00149785
InChI Key WRUGWIBCXHJTDG-UHFFFAOYSA-L
Synonym magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
PubChem CID 24843
ChEBI CHEBI:31795
IUPAC Name magnesium;sulfate;heptahydrate
SMILES O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

Specifications

Melting Point 770°C
Color White
pH 5 to 8.2
Physical Form Solid
Quantity 1 kg
Packaging Plastic powder jar
Vapor Pressure <0.1mmHg at 20°C
Formula Weight 246.48g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.