missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Sulfate Heptahydrate, Fisher BioReagents

Product Code. 10553335
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Poly Bottle
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10553335 1 kg Poly Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10553335 Supplier Fisher BioReagents Supplier No. BP2131
Spotlight Offers
Use promo code 28036
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
This item is not returnable. View return policy

CAS: 10034-99-8 | H14MgO11S | 246.47 g/mol

Poly Bottle; 1kg

Magnesium Sulfate is suitable for use in the preparation of buffer solutions.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Magnesium Sulfate Heptahydrate
Melting Point 770°C
CAS 10034-99-8
Color White
pH 5 to 8.2
Physical Form Solid
Quantity 1 kg
Assay Percent Range 98.0 to 102.0 %
Molecular Formula H14MgO11S
MDL Number MFCD00149785
Merck Index 15, 5755
Synonym magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
InChI Key WRUGWIBCXHJTDG-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O
IUPAC Name magnesium;sulfate;heptahydrate
Molecular Weight (g/mol) 246.47
PubChem CID 24843
ChEBI CHEBI:31795
Formula Weight 246.47
Percent Purity 98.0 to 102.0%
Identification Pass Test
Packaging Poly Bottle
Vapor Pressure <0.1 mmHg at 20°C
Density 1.678g/cm³
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.