missing translation for 'onlineSavingsMsg'
Learn More

Magnesium sulfate heptahydrate, puriss. p.a., ACS reagent, Solstice

Product Code. 15676080
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15676080 100 g 100g
15696080 500 g 500g
15686080 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15676080 Supplier Honeywell-Fluka Supplier No. 63140100G
  • ONLINE EXCLUSIVE PRICE
    36.31 €
    32.83 € / 100g
    Save 3.48 €
    9% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-99-8
Molecular Formula H14MgO11S
Molecular Weight (g/mol) 246.47
MDL Number MFCD00149785
InChI Key WRUGWIBCXHJTDG-UHFFFAOYSA-L
Synonym magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
PubChem CID 24843
ChEBI CHEBI:31795
IUPAC Name magnesium;sulfate;heptahydrate
SMILES O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

Specifications

Physical Form Crystals
Quantity 100 g
Vapor Pressure 0.1 mmHg (20°C)
Density Vapor Density: <0.01 (vs air)
Linear Formula MgSO4 · 7H2O
UN Number NONH for all modes of transport
Formula Weight 246.47g/mol
Percent Purity ≥99% (KT)
Grade ACS/puriss. p.a.
Chemical Name or Material Magnesium sulfate heptahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.