missing translation for 'onlineSavingsMsg'
Learn More

Magnesium sulfate hydrate, Puriss., meets analytical spec. of DAC, dried, 99.0-101.0% MgSO4 basis (in dried substance), Solstice
BIOPHARMA

Product Code. 15611260 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
5 kg
25 kg
6 x 1 kg
6 x 2.5 kg
Unit Size:
1kg
2.5kg
25kg
5kg
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15611260 1 kg 1kg
15621260 2.5 kg 2.5kg
15641260 5 kg 5kg
15651260 6 x 1 kg Pack of 6
15661260 6 x 2.5 kg Pack of 6
15631260 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15611260 Supplier Honeywell-Fluka Supplier No. 131431KG
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
This item is not returnable. View return policy

Puriss., meets analytical specification of DAC, dried, 99.0-101.0% MgSO4 basis (in dried substance)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22189-08-8
Molecular Formula MgO4S
Molecular Weight (g/mol) 120.36
MDL Number MFCD00149787
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
Synonym magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
PubChem CID 24083
ChEBI CHEBI:32599
SMILES [Mg++].[O-]S([O-])(=O)=O

Specifications

Physical Form Dry
Quantity 1 kg
Linear Formula MgSO4 · aq
UN Number NONH for all modes of transport
IUPAC Name magnesium;sulfate
Formula Weight 120.37g/mol (anhydrous basis)
Percent Purity 99 to 101% (MgSO4 basis) (in dried substance)
Grade puriss.
Chemical Name or Material Magnesium Sulfate Hydrate, Meets analytical specification of DAC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.