missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Sulfate Solution, 0.1M, Solstice

Product Code. 15694140
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15694140 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15694140 Supplier Honeywell-Fluka Supplier No. 342761L
  • ONLINE EXCLUSIVE PRICE
    149.00 €
    134.37 € / 1 L
    Save 14.63 €
    9% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Volumetric, 0.1 M MgSO4

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Specifications

CAS 7487-88-9
Quantity 1 L
Molecular Formula MgO4S
MDL Number MFCD00011110
UN Number NONH for all modes of transport
Synonym magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
SMILES [Mg++].[O-]S([O-])(=O)=O
IUPAC Name magnesium;sulfate
Molecular Weight (g/mol) 120.36
PubChem CID 24083
ChEBI CHEBI:32599
Formula Weight 120.37g/mol
Chemical Name or Material Magnesium sulfate solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.