missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Sulfate, TRC
Specifications
Specifications
| Chemical Name or Material | Magnesium Sulfate |
| CAS | 7487-88-9 |
| Quantity | 10 g |
| Molecular Formula | MgSO4 |
| Synonym | Sulfuric acid magnesium salt (1:1) (8CI, 9CI, ACI), Magnesium sulfate (1:1) (ACI), Arrosalt 2327, Epso Top, Epsom salt, Epsom salts, M-HOS, Magnesii sulfas, Magnesium sulfate, Magnesium sulphate, MG 3K, MG-N, Mg-OK, MG-OK 100, MN 00, MN 30, MN 70, NP-YW 1, OT-S, OT-S (drying agent), SN 00, SSN 00, Sulfuric acid magnesium salt, Sulfuric acid, magnesium salt (1:1), Sunmate, TC-P, Tomix OT, USN 00, WS 1S |
| InChI Formula | InChI=1S/Mg.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2 |
| SMILES | [Mg+2].[O-]S(=O)(=O)[O-] |
| IUPAC Name | magnesium;sulfate |
| Molecular Weight (g/mol) | 120.37 |
| Formula Weight | 119.9368 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
¿Detecta una oportunidad de mejora?