missing translation for 'onlineSavingsMsg'
Learn More

Magnesium trifluoromethanesulfonate, 97%

Product Code. 11458217
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11458217 100 g 100g
11438217 5 g 5g
11448217 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11458217 Supplier Thermo Scientific Alfa Aesar Supplier No. A17591.22
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 60871-83-2 | C2F6MgO6S2 | 322.431 g/mol

Magnesium trifluoromethanesulfonate is used as a component of the electrolyte, as lewis acid resulted in largely decreased and reversed enantioselectivities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium trifluoromethanesulfonate is used as a component of the electrolyte, as lewis acid resulted in largely decreased and reversed enantioselectivities.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60871-83-2
Molecular Formula C2F6MgO6S2
Molecular Weight (g/mol) 322.431
MDL Number MFCD00042601
InChI Key BZQRBEVTLZHKEA-UHFFFAOYSA-L
Synonym magnesium trifluoromethanesulfonate, magnesium triflate, magnesium 2+ ditriflate, trifluoromethanesulfonic acid magnesium salt, magnesium trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, magnesium salt, magnesiumtrifluoromethanesulfonate, magnesium trifiate, acmc-209mmv, ksc352a4p
PubChem CID 2734125
IUPAC Name magnesium;trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Mg+2]

Specifica

Melting Point >310°C
Linear Formula Mg(CF3SO3)2
Quantity 100 g
UN Number UN1759
Beilstein 705578
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 322.43
Percent Purity 98%
Chemical Name or Material Magnesium trifluoromethanesulfonate

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.