Learn More
Manutan Magnetic easy wipe racking strip, yellow, 10m x 50mm
Description
Cut with scissors to provide an easy, flexible solution to location identification. 10m long plasticised magnetic marker strips. Write on and wipe off using standard wet wipe marker pens. Ideal for cold stores.
Roll width : 50mm
Roll width : 50mm