missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Magnetic Plastic Transport Signs

Product Code. 11714904 Shop All Brady Products
Change view
Click to view available options
Displayed Text:
Corrosive Substance 8
Environmentally Hazardous Substances
Miscellaneous hazardous substances 9
Non-flammable, non-toxic gas 2
Organic Peroxide 5.2
Oxidizing Substance 5.1
Toxic and Harmful Substances 6
Shape:
Diamond
Rhomboid
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Displayed Text Shape Unit Size
11714904 Non-flammable, non-toxic gas 2 Rhomboid Each
11774914 Corrosive Substance 8 Diamond Each
11794914 Miscellaneous hazardous substances 9 Diamond Each
11730244 Non-flammable, non-toxic gas 2 Diamond Each
11750953 Organic Peroxide 5.2 Diamond Each
11714914 Oxidizing Substance 5.1 Diamond Each
11734914 Toxic and Harmful Substances 6 Diamond Each
11721003 Environmentally Hazardous Substances Rhomboid Each
11700963 Organic Peroxide 5.2 Rhomboid Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 11714904 Supplier Brady™ Supplier No. 258943
  • ONLINE EXCLUSIVE PRICE
    25.65 €
    24.48 € / Pack of 1
    Save 1.17 €
    4% Off
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Certifications/Compliance ISO 7010
Alternate Part Number ADR 2.2B-297*297-B0859
Material Plastic
Quantity 1 Ea.
Adhesive Type Permanent Acrylic
Description Magnetic Plastic
Height (Metric) 297 mm
Width (Metric) 297 mm
Temperature Range (Metric) -18°C to 70°C
Shape Rhomboid
Displayed Text Non-flammable, non-toxic gas 2
Product Type Transport Sign
Color Black on Green
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.