missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Can be used for the purification of strep-tagged fusion proteins in batch in single or high-throughput format.
- Beads show low non-specific protein binding due to their agarose surface and high specificity of Strep-Tactin™ XT towards Strep-tag. This results in outstanding target protein purity and makes the MagStrep beads a perfect tool for the isolation of proteins or the identification of complexes via pull-downs, immunoprecipitation (IP), or co-immunoprecipitation (Co-IP).
- MagStrep Strep-Tactin™ XT beads are suitable for fast small-scale purifications in reaction tubes as well as 96-well plates.
- The beads can be easily and swiftly separated with a magnetic separator. This eliminates the need for centrifugation and reduces the risk of disturbing the pellet or loss of beads within the wash step to zero.
- The elution of target proteins or complexes can be triggered under mild conditions with the biotin-containing Buffer BXT or under denaturing conditions by boiling in SDS gel loading buffer.
Specifications
Specifications
| Formulation | 5% (v/v) Suspension in PBS with 0.1% (w/v) Sodium Azide |
| Support Type | 6% Agarose, Crosslinked |
| Particle Size | 20 to 40 μm |
| Form | Resin Slurry |
| Target | Twin-Strep-tag or Strep-tag II |
| Type | Magnetic Agarose Bead |
| Capacity (Metric) | 10 mL |
For research use only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?