missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences MagStrep Strep-Tactin™ XT Beads

Product Code. 17942451 Shop All IBA Lifesciences Products
Change view
Click to view available options
Capacity (Metric):
2 mL
10 mL
Unit Size:
10mL
2mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
17942451 2 mL 2mL
17932451 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17942451 Supplier IBA Lifesciences Supplier No. 25090002
  • ONLINE EXCLUSIVE PRICE
    238.00 €
    209.00 € / 2mL
    Save 29.00 €
    12% Off
 Restricted Item
In Stock
Ajouter au panier
Edge
Ajouter au panier

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

MagStrep Strep-Tactin™ XT beads are magnetic beads (5% suspension) to purify Strep-tag™ II or Twin-Strep-tag™ fusion proteins.

  • Can be used for the purification of strep-tagged fusion proteins in batch in single or high-throughput format.
  • Beads show low non-specific protein binding due to their agarose surface and high specificity of Strep-Tactin™ XT towards Strep-tag. This results in outstanding target protein purity and makes the MagStrep beads a perfect tool for the isolation of proteins or the identification of complexes via pull-downs, immunoprecipitation (IP), or co-immunoprecipitation (Co-IP).
  • MagStrep Strep-Tactin™ XT beads are suitable for fast small-scale purifications in reaction tubes as well as 96-well plates.
  • The beads can be easily and swiftly separated with a magnetic separator. This eliminates the need for centrifugation and reduces the risk of disturbing the pellet or loss of beads within the wash step to zero.
  • The elution of target proteins or complexes can be triggered under mild conditions with the biotin-containing Buffer BXT or under denaturing conditions by boiling in SDS gel loading buffer.

Specifications

Formulation 5% (v/v) Suspension in PBS with 0.1% (w/v) Sodium Azide
Support Type 6% Agarose, Crosslinked
Particle Size 20 to 40 μm
Form Resin Slurry
Target Twin-Strep-tag or Strep-tag II
Type Magnetic Agarose Bead
Capacity (Metric) 2 mL

For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.