missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Malachite Green-d5 Oxalate (~85%), TRC
Specifications
Specifications
| Chemical Name or Material | Malachite Green-d5 Oxalate |
| Molecular Formula | C23H20D5N2+ • C2HO4- |
| Quantity | 2.5 mg |
| Synonym | N-[4-[[4-(Dimethylamino)phenyl]phenylmethylene]-2,5-cyclohexadien-1-ylidene]-N-methylmethanaminium-d5 Oxalate, Aniline Green-d5, Astra Malachite Gree-d5n, Astra Malachite Green B-d5, Astra Malachite Green BXXv, Basic Green 4-d5, Basonyl Green 830-d5, Basonyl Green NB 832-d5, Benzal Green Oxalate-d5, Victoria Green Oxalate-d5 |
| InChI Formula | InChI=1S/C23H25N2.C2H2O4/c1-24(2)21-14-10-19(11-15-21)23(18-8-6-5-7-9-18)20-12-16-22(17-13-20)25(3)4;3-1(4)2(5)6/h5-17H,1-4H3;(H,3,4)(H,5,6)/q+1;/p-1/i5D,6D,7D,8D,9D; |
| SMILES | O=C(C(O)=O)[O-].[2H]C(C([2H])=C1[2H])=C([2H])C([2H])=C1/C(C2=CC=C(N(C)C)C=C2)=C3C=C/C(C=C/3)=[N+](C)\C |
| Molecular Weight (g/mol) | 334.498903 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?