missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Malachite Green (Oxalate), Pure, C.I.42000, Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Melting Point | 164°C |
| CAS | 2437-29-8 |
| Molecular Formula | C52H54N4O12 |
| MDL Number | MFCD00011766,MFCD00151209 |
| InChI Key | CNYGFPPAGUCRIC-UHFFFAOYSA-L |
| SMILES | OC(=O)C(O)=O.OC(=O)C([O-])=O.OC(=O)C([O-])=O.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C |
| IUPAC Name | [4-[[4-(dimethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;2-hydroxy-2-oxoacetate;oxalic acid |
| Molecular Weight (g/mol) | 927.02 |
| PubChem CID | 2724411 |
| Formula Weight | 927.03g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction