missing translation for 'onlineSavingsMsg'
Learn More

Malachite green oxalate salt, MP Biomedicals™

Product Code. 11487982 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
Unit Size:
25g

Product Code. 11487982

Brand: MP Biomedicals 0215264225

Please to purchase this item. Need a web account? Register with us today!

Specifications

CAS 2437-29-8
Molecular Formula C52H54N4O12
MDL Number MFCD00011766,MFCD00151209
Synonym N,N,N',N'-Tetramethyl-4,4'-diaminotriphenylcarbenium oxalate, N, N, N'
InChI Key CNYGFPPAGUCRIC-UHFFFAOYSA-L
SMILES OC(=O)C(O)=O.OC(=O)C([O-])=O.OC(=O)C([O-])=O.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C
IUPAC Name bis(bis[4-(dimethylamino)phenyl](phenyl)methylium) oxalic acid dihydrogen oxalate
Molecular Weight (g/mol) 927.02
PubChem CID 2724411
Formula Weight 929
Color Green
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Malachite green oxalate salt
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.