Learn More
Value Plastics Male Luer Integral Lock Ring to Female Luer Thread Style Coupler
Shop All Cole-Parmer ProductsDescription
Luer fittings allow for a simple yet secure twist-lock connection. Easily transition from a male luer to female luer fitting. Ideal for general laboratory and single-use applications
Specifications
Specifications
| Certifications/Compliance | Non-DEHP;Non-Phthalate;RoHS |
| Inlet | Male Luer Integral Lock Ring, Female Luer Thread |
| Material (Seal) | Nylon |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.