Learn More
Thermo Scientific™ Male Luer-LOK Priming Syringes
Thermo Scientific™ Male Luer-LOK Priming Syringes offer easy, accurate and reproducible manual injections.
66.61 € - 363.65 €
Product Code | Brand | Volume (Metric) | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Product Type | Price | Quantity & Availability | ||||
10524655
|
Thermo Scientific™
365KL531 |
1 mL | Gas Tight GC Autosampler Syringe, Luer-Lok |
91.42 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13436886
|
Thermo Scientific™
365N2721 |
10 mL | GC Autosampler Syringe |
363.65 €
Each |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10409254
|
Thermo Scientific™
365LL541 |
2.5 mL | Gas Tight GC Autosampler Syringe, Luer-Lok |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10137854
|
Thermo Scientific™
365ML551 |
5 mL | Gas Tight GC Autosampler Syringe, Luer-Lok |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10585995
|
Thermo Scientific™
365NL561 |
10 mL | Gas Tight GC Autosampler Syringe, Luer-Lok |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10409444
|
Thermo Scientific™
365PL571 |
25 mL | Gas Tight GC Autosampler Syringe, Luer-Lok |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10351305
|
Thermo Scientific™
365RL581 |
50 mL | GC Autosampler Priming Syringe, Luer-Lok |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Thermo Scientific™ Male Luer-LOK Priming Syringes offer easy, accurate and reproducible manual injections.
- Square tip to prevent damage to the injector
- Wide range of volumes
- Precision made from borosilicate glass and stainless steel
- Robust design and easy-to-read markings
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.