missing translation for 'onlineSavingsMsg'
Learn More

Maleic Acid Monomethyl Ester (~90%), TRC

Product Code. 30478042
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478042 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30478042 Supplier Toronto Research Chemicals Supplier No. TRCM124395100G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Maleic Acid Monomethyl Ester
CAS 3052-50-4
Molecular Formula C5H6O4
Quantity 100 g
Synonym (2Z)-2-Butenedioic Acid 1-Methyl Ester, (2Z)-2-Butenedioic Acid Monomethyl Ester, Maleic Acid Methyl Ester, (Z)-4-Methoxy-4-oxobut-2-enoic Acid, (Z)-4-Methoxy-4-oxobut-2-enoic Acid, Hydrogen Methyl Maleate, Methyl Hydrogen (2Z)-2-Butendioate, Methyl Hydrogen Maleate, Monomethyl (Z)-2-Butenedioate, Monomethyl Maleate
InChI Formula InChI=1S/C5H6O4/c1-9-5(8)3-2-4(6)7/h2-3H,1H3,(H,6,7)/b3-2-
SMILES COC(=O)\C=C/C(=O)O
IUPAC Name (Z)-4-methoxy-4-oxobut-2-enoic acid
Molecular Weight (g/mol) 130.1
Formula Weight 130.0266
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.