missing translation for 'onlineSavingsMsg'
Learn More

Malic acid, MedChemExpress

Product Code. 16412728
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16412728 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16412728 Supplier Medchem Express Supplier No. HYY1311
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Malic acid (Hydroxybutanedioic acid) is a dicarboxylic acid that is naturally found in fruits such as apples and pears. It plays a role in many sour or tart foods.

Malic acid (Hydroxybutanedioic acid) is a dicarboxylic acid that is naturally found in fruits such as apples and pears. It plays a role in many sour or tart foods.

Specifications

Chemical Name or Material Malic acid
CAS 6915-15-7
Color White
Purity Grade Notes Research
Molecular Formula C4H6O5
Quantity 5 g
Synonym Hydroxybutanedioic acidE 296
Solubility Information DMSO : 100 mg/mL (745.77 mM; Need ultrasonic) ∣H2O : 100 mg/mL (745.77 mM; Need ultrasonic)
SMILES O=C(O)C(O)CC(O)=O
Molecular Weight (g/mol) 134.09
Formula Weight 134.09
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.