missing translation for 'onlineSavingsMsg'
Learn More

Malonaldehyde bis(phenylimine) monohydrochloride, 97+%

Product Code. 11460327
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460327 25 g 25g
11470327 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11460327 Supplier Thermo Scientific Alfa Aesar Supplier No. B21721.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 123071-42-1 | C15H15ClN2 | 258.749 g/mol

Malonaldehyde bis(phenylimine) monohydrochloride is a useful fluorescent cyanine dye.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Malonaldehyde bis(phenylimine) monohydrochloride is a useful fluorescent cyanine dye.

Solubility
Soluble in water (partly).

Notes
Hygroscopic, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123071-42-1
Molecular Formula C15H15ClN2
Molecular Weight (g/mol) 258.749
MDL Number MFCD00054634
InChI Key FLMLOZWXXPLHIE-UHFFFAOYSA-N
Synonym malonaldehyde bis phenylimine monohydrochloride, malonaldehyde bis phenylimine hydrochloride, malondialdehyde bis phenylimine monohydrochloride, malonaldehyde bis phenylimine hcl, malonaldehydedianilidehydrochloride, n-3-phenylimino-1-propen-1-yl aniline hydrochloride, n1,n3-diphenylpropane-1,3-diimine hydrochloride, 1e,3e-n1,n3-diphenylpropane-1,3-diimine hydrochloride
PubChem CID 12213307
IUPAC Name N,N'-diphenylpropane-1,3-diimine;hydrochloride
SMILES C1=CC=C(C=C1)N=CCC=NC2=CC=CC=C2.Cl

Specifications

Melting Point ∽215°C (decomposition)
Quantity 25 g
Solubility Information Soluble in water (partly).
Formula Weight 258.75
Percent Purity 98%
Chemical Name or Material Malonaldehyde bis(phenylimine) monohydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.