missing translation for 'onlineSavingsMsg'
Learn More

Malonamide, 98%

Product Code. 11459066
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459066 100 g 100g
11469066 500 g 500g
11479066 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11459066 Supplier Thermo Scientific Alfa Aesar Supplier No. A17939.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 108-13-4 | C3H6N2O2 | 102.093 g/mol

Malonamide is a reagent for fluorimetric determination of reducing carbohydrates. The malonamide-based ionic liquid extractant was used in the extraction of europium(iii) and other trivalent rare earth elements from nitric acid medium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Malonamide is a reagent for fluorimetric determination of reducing carbohydrates. The malonamide-based ionic liquid extractant was used in the extraction of europium(iii) and other trivalent rare earth elements from nitric acid medium.

Solubility
Soluble in HCl.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-13-4
Molecular Formula C3H6N2O2
Molecular Weight (g/mol) 102.093
MDL Number MFCD00008034
InChI Key WRIRWRKPLXCTFD-UHFFFAOYSA-N
Synonym malonamide, malonodiamide, malondiamide, malonyldiamide, carboxamidoacetamide, malonic acid diamide, malonic diamide, unii-qvq8cng255, qvq8cng255, methane-1,1-dicarboxamide
PubChem CID 7911
IUPAC Name propanediamide
SMILES C(C(=O)N)C(=O)N

Specifications

Melting Point 167°C to 171°C
Quantity 100 g
Beilstein 1751401
Solubility Information Soluble in HCl.
Formula Weight 102.09
Percent Purity 98%
Chemical Name or Material Malonamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.