missing translation for 'onlineSavingsMsg'
Learn More

Malondialdehyde Bis(phenylimine) Monohydrochloride, TRC

Product Code. 30466515
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30466515 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30466515 Supplier Toronto Research Chemicals Supplier No. TRCM198140250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Malondialdehyde Bis(phenylimine) Monohydrochloride
CAS 123071-42-1
Molecular Formula C15H14N2 . HCl
Quantity 250 mg
Synonym Benzenamine, N,N'-1,3-propanediylidenebis-, hydrochloride (1:1) (ACI), Benzenamine, N,N'-1,3-propanediylidenebis-, monohydrochloride (9CI), Malonaldehyde bis(phenylimine) monohydrochloride, Malonaldehyde dianilide hydrochloride, Malondialdehyde bis(phenylimine) monohydrochloride, N,N'-1,3-Propanediylidenebis[benzenamine] hydrochloride (1:1), Malonaldehyde dianilide monohydrochloride
InChI Formula InChI=1S/C15H14N2.ClH/c1-3-8-14(9-4-1)16-12-7-13-17-15-10-5-2-6-11-15;/h1-6,8-13H,7H2;1H
SMILES Cl.C(C=Nc1ccccc1)C=Nc2ccccc2
IUPAC Name N,N'-diphenylpropane-1,3-diimine;hydrochloride
Molecular Weight (g/mol) 258.75
Formula Weight 258.0924
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.