missing translation for 'onlineSavingsMsg'
Learn More

Malonic Acid Disodium Salt Monohydrate, TRC

Product Code. 30482209
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30482209 50 g 50g
30480856 10 g 10g
30471420 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30482209 Supplier Toronto Research Chemicals Supplier No. TRCM15803050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26522-85-0
Molecular Formula C3H2Na2O4 . H2O
Molecular Weight (g/mol) 166.04
IUPAC Name disodium;propanedioate;hydrate
SMILES O.[Na+].[Na+].[O-]C(=O)CC(=O)[O-]

Specifications

Quantity 50 g
InChI Formula InChI=1S/C3H4O4.2Na.H2O/c4-2(5)1-3(6)7;;;/h1H2,(H,4,5)(H,6,7);;;1H2/q;2*+1;/p-2
Formula Weight 165.9854
Chemical Name or Material Malonic Acid Disodium Salt Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.