missing translation for 'onlineSavingsMsg'
Learn More

Malonic Acid, TRC

Product Code. 30006093
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30006093 100 g 100g
30005980 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30006093 Supplier Toronto Research Chemicals Supplier No. TRCM158005100G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-82-2
Molecular Formula C3 H4 O4
Molecular Weight (g/mol) 104.06
Synonym Propanedioic acid (9CI, ACI), Malonic acid (8CI), 1,3-Propanedioic acid, Carboxyacetic acid, Dicarboxymethane, Malonates, MeSH ID: D008314, Methanedicarboxylic acid, NSC 8124
IUPAC Name propanedioic acid
SMILES OC(=O)CC(=O)O

Specifications

Quantity 100 g
InChI Formula InChI=1S/C3H4O4/c4-2(5)1-3(6)7/h1H2,(H,4,5)(H,6,7)
Formula Weight 104.011
Chemical Name or Material Malonic acid (Propanedioic acid)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.