missing translation for 'onlineSavingsMsg'
Learn More

Malononitrile, 99%

Product Code. 11477307
Click to view available options
Quantity:
5 g
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
5g
This item is not returnable. View return policy

Product Code. 11477307

Brand: Thermo Scientific Alfa Aesar A15046.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-77-3 | C3H2N2 | 66.063 g/mol

Crosslinking agent for modification of proteins via amidationMalononitrile is used as a precursor in the preparation of anti-cancer drug, triamterene, adenine, methotrexate, thiamine, acrylic fibers and dyes. It is used in Knoevenagel condensation to prepare 2-chlorobenzalmalononitrile by reacting with 2-chlorobenzaldehyde. It acts as a leaching agent for gold. It is used as a raw material for the Gewald reaction of ketone or aldehyde to produce 2-aminothiophene using elemental sulfur and a base. Further, it plays an important role in the preparation of [1,6]-naphthyridines, gamma-ketoamides, 1,4-dihydropyridine, chromeno[2,3-b]pyridine and dihydro-1,4-dithiepine frameworks.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Crosslinking agent for modification of proteins via amidationMalononitrile is used as a precursor in the preparation of anti-cancer drug, triamterene, adenine, methotrexate, thiamine, acrylic fibers and dyes. It is used in Knoevenagel condensation to prepare 2-chlorobenzalmalononitrile by reacting with 2-chlorobenzaldehyde. It acts as a leaching agent for gold. It is used as a raw material for the Gewald reaction of ketone or aldehyde to produce 2-aminothiophene using elemental sulfur and a base. Further, it plays an important role in the preparation of [1,6]-naphthyridines, gamma-ketoamides, 1,4-dihydropyridine, chromeno[2,3-b]pyridine and dihydro-1,4-dithiepine frameworks.

Solubility
Soluble in water, acetone, acetic acid, chloroform, ethanol, ether and benzene.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong reducing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-77-3
Molecular Formula C3H2N2
MDL Number MFCD00001883
InChI Key CUONGYYJJVDODC-UHFFFAOYSA-N
Synonym malononitrile, dicyanomethane, cyanoacetonitrile, malonic dinitrile, methylene cyanide, dicyanmethane, malonodinitrile, malonic acid dinitrile, methylenedinitrile, methane, dicyano
PubChem CID 8010
ChEBI CHEBI:33186
IUPAC Name propanedinitrile

Specifications

Melting Point 30°C to 34°C
Density 1.049
Boiling Point 220°C to 222°C
Flash Point 112°C (233°F)
Refractive Index 1.415
Quantity 500 g
UN Number UN2647
Beilstein 773697
Merck Index 14,5711
Solubility Information Soluble in water,acetone,acetic acid,chloroform,ethanol,ether and benzene.
SMILES C(C#N)C#N
Molecular Weight (g/mol) 66.063
Formula Weight 66.06
Percent Purity 99%
Chemical Name or Material Malononitrile
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.