missing translation for 'onlineSavingsMsg'
Learn More

Malononitrile, 99%

Product Code. 11487307
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11487307 2500 g 2500g
11408840 5 g 5g
11467307 100 g 100g
11477307 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11487307 Supplier Thermo Scientific Alfa Aesar Supplier No. A15046.0E
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 109-77-3 | C3H2N2 | 66.063 g/mol

Crosslinking agent for modification of proteins via amidationMalononitrile is used as a precursor in the preparation of anti-cancer drug, triamterene, adenine, methotrexate, thiamine, acrylic fibers and dyes. It is used in Knoevenagel condensation to prepare 2-chlorobenzalmalononitrile by reacting with 2-chlorobenzaldehyde. It acts as a leaching agent for gold. It is used as a raw material for the Gewald reaction of ketone or aldehyde to produce 2-aminothiophene using elemental sulfur and a base. Further, it plays an important role in the preparation of [1,6]-naphthyridines, gamma-ketoamides, 1,4-dihydropyridine, chromeno[2,3-b]pyridine and dihydro-1,4-dithiepine frameworks.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Crosslinking agent for modification of proteins via amidationMalononitrile is used as a precursor in the preparation of anti-cancer drug, triamterene, adenine, methotrexate, thiamine, acrylic fibers and dyes. It is used in Knoevenagel condensation to prepare 2-chlorobenzalmalononitrile by reacting with 2-chlorobenzaldehyde. It acts as a leaching agent for gold. It is used as a raw material for the Gewald reaction of ketone or aldehyde to produce 2-aminothiophene using elemental sulfur and a base. Further, it plays an important role in the preparation of [1,6]-naphthyridines, gamma-ketoamides, 1,4-dihydropyridine, chromeno[2,3-b]pyridine and dihydro-1,4-dithiepine frameworks.

Solubility
Soluble in water, acetone, acetic acid, chloroform, ethanol, ether and benzene.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong reducing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-77-3
Molecular Formula C3H2N2
MDL Number MFCD00001883
InChI Key CUONGYYJJVDODC-UHFFFAOYSA-N
Synonym malononitrile, dicyanomethane, cyanoacetonitrile, malonic dinitrile, methylene cyanide, dicyanmethane, malonodinitrile, malonic acid dinitrile, methylenedinitrile, methane, dicyano
PubChem CID 8010
ChEBI CHEBI:33186
IUPAC Name propanedinitrile

Specifications

Melting Point 30°C to 34°C
Density 1.049
Boiling Point 220°C to 222°C
Flash Point 112°C (233°F)
Refractive Index 1.415
Quantity 2500 g
UN Number UN2647
Beilstein 773697
Merck Index 14,5711
Solubility Information Soluble in water,acetone,acetic acid,chloroform,ethanol,ether and benzene.
SMILES C(C#N)C#N
Molecular Weight (g/mol) 66.063
Formula Weight 66.06
Percent Purity 99%
Chemical Name or Material Malononitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.