missing translation for 'onlineSavingsMsg'
Learn More

Malononitrile, TRC

Product Code. 30481825
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481825 100 g 100g
30474946 25 g 25g
30467435 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30481825 Supplier Toronto Research Chemicals Supplier No. TRCM158020100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-77-3
Molecular Formula C3 H2 N2
Molecular Weight (g/mol) 66.06
Synonym Propanedinitrile, Malononitrile (8CI), Cyanoacetonitrile, Dicyanomethane, Malondinitrile, Malonic acid dinitrile, Malonodinitrile, Methane, dicyano-, Methylene cyanide, Methylenedinitrile, NSC 3769, Malonitrile
IUPAC Name propanedinitrile
SMILES N#CCC#N

Specifications

Quantity 100 g
InChI Formula InChI=1S/C3H2N2/c4-2-1-3-5/h1H2
Formula Weight 66.0218
Chemical Name or Material Malonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.