missing translation for 'onlineSavingsMsg'
Learn More

Malonyl dichloride, 97%

Product Code. 10699504 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10699504 25 g Glass bottle 25g
10692135 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10699504 Supplier Thermo Scientific Acros Supplier No. 125280250
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1663-67-8 | C3H2Cl2O2 | 140.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1663-67-8
Molecular Formula C3H2Cl2O2
Molecular Weight (g/mol) 140.95
MDL Number MFCD00000735
InChI Key SXYFKXOFMCIXQW-UHFFFAOYSA-N
Synonym malonyl chloride, malonyl dichloride, malonoyl chloride, malonic acid dichloride, malonoyl dichloride, malonic acid chloride, malonylchloride, methane-1,1-dicarbonyl chloride, malonylchlorid, malonyldichloride
PubChem CID 74269
IUPAC Name propanedioyl dichloride
SMILES C(C(=O)Cl)C(=O)Cl

Specifications

Color Brown to Yellow
Density 1.4500g/mL
Boiling Point 47.0°C (15.0 mmHg)
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Linear Formula ClCOCH2COCl
Refractive Index 1.4620 to 1.4660
Quantity 25 g
Beilstein 02, I, 252
Fieser 12,291
Specific Gravity 1.45
Solubility Information Solubility in water: reacts violently. Other solubilities: soluble in ether
Formula Weight 140.95
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Malonyl dichloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Malonyl dichloride
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.