missing translation for 'onlineSavingsMsg'
Learn More

Melford Malt Extract Agar

Product Code. 30018459
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
This item is not returnable. View return policy

Product Code. 30018459

Brand: Melford M21500500.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Malt Extract Agar is used for the isolation, cultivation and maintenance of yeasts and molds.

Malt Extract Agar is used for the isolation, cultivation and maintenance of yeasts and molds. Formulation is similar to Malt Extract Broth with the addition of agar and glycerol. This broth medium was recommended by Thom and Church for the cultivation of fungi. Its low acidity inhibits the growth of most bacteria. The high carbohydrate content enhances the rapid growth of fungi and yeasts.

Dissolve 33.6 grams in 1 liter of distilled water. Sterilize by autoclaving for 15 minutes.

Ingredients:

Maltose: 12.75 g/L

Dextrin: 2.75 g/L

Glycerol: 2.35 g/L

Peptone: 0.78 g/L

Agar: 15.0 g/L

Final pH (25°C): 4.6 ± 0.2

Specifications

Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.