missing translation for 'onlineSavingsMsg'
Learn More

Maltitol, 97%

Product Code. 11441177
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11441177 250 g 250g
11431177 50 g 50g
11421177 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11441177 Supplier Thermo Scientific Alfa Aesar Supplier No. B24158.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 585-88-6 | C12H24O11 | 344.313 g/mol

Maltitol is used in the manufacturing of sweets, including sugarless hard candies, chewing gum, chocolates, baked goods and ice cream. In the pharmaceutical industry, it is used as an excipient. Further, it is used as a plasticizer in gelatin capsules, humectant, emollient and a low-calorie sweetening agent. It is involved in the study to investigate the comparative effects of the maltitol chewing gums on reducing plaque.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Maltitol is used in the manufacturing of sweets, including sugarless hard candies, chewing gum, chocolates, baked goods and ice cream. In the pharmaceutical industry, it is used as an excipient. Further, it is used as a plasticizer in gelatin capsules, humectant, emollient and a low-calorie sweetening agent. It is involved in the study to investigate the comparative effects of the maltitol chewing gums on reducing plaque.

Solubility
Soluble in water. Slightly soluble in ethanol.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 585-88-6
Molecular Formula C12H24O11
Molecular Weight (g/mol) 344.313
MDL Number MFCD00006600
InChI Key VQHSOMBJVWLPSR-WUJBLJFYSA-N
Synonym maltitol, d-maltitol, maltisorb, malbit, amalti syrup, malti mr, amalty mr 100, amalty, 4-o-alpha-d-glucopyranosyl-d-glucitol, unii-d65dg142wk
PubChem CID 493591
ChEBI CHEBI:68428
IUPAC Name (2S,3R,4R,5R)-4-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexane-1,2,3,5,6-pentol
SMILES C(C1C(C(C(C(O1)OC(C(CO)O)C(C(CO)O)O)O)O)O)O

Specifications

Melting Point 149°C to 152°C
Assay Percent Range 0.97
Quantity 250 g
Beilstein 89983
Solubility Information Soluble in water. Slightly soluble in ethanol.
Formula Weight 344.32
Percent Purity 97%
Chemical Name or Material Maltitol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.