missing translation for 'onlineSavingsMsg'
Learn More

Maltotriose, 93%

Product Code. 10691555 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
25 g
Unit Size:
25g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10691555 500 mg 500mg
10127843 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10691555 Supplier Thermo Scientific Acros Supplier No. 225945000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1109-28-0 | C18H32O16 | 504.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1109-28-0
MDL Number MFCD00006629
InChI Key FYGDTMLNYKFZSV-NBCPLHMPSA-N
Synonym maltotriose, d-maltotriose
PubChem CID 134129496
IUPAC Name (2R,3S,4S,5S,6R)-2-[(2R,3R,4R,5R,6R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(2R,3S,4S,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)O)O

Specifications

Melting Point 132.0°C to 135.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 91% min. (ELSD) (HPLC)
Packaging Glass bottle
Molecular Formula C18H32O16
Quantity 500 mg
Specific Rotation Condition +162.50° (25°C c=1,H2O)
Specific Rotation + 162.50
Solubility Information Solubility in water: 50mg/mL,clear colorless solution
Molecular Weight (g/mol) 504.44
Formula Weight 504.44
Percent Purity 93%
Physical Form Fine Crystalline Powder or Crystals
Chemical Name or Material Maltotriose
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.