missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences pCSG-IBA Mammalian Expression Vector with No Secretion
Designed for high-level episomal expression in mammalian hosts under G418 selection. Mammalian Expression Vector with No Secretion is a large expression vector with universal features for transient expression as well as for generation of stable cell lines (NeoR).
220.00 € - 266.00 €
Specifications
Constitutive or Inducible System | Constitutive |
---|---|
Marker Gene | Ampicillin |
Promoter | CMV |
Product Type | Vector |
Selection Agent (Eukaryotic) | Geneticin™ (G-418) |
Product Code | Brand | Protein Tag | Vector | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Protein Tag | Vector | Price | Quantity & Availability | ||||
15613858
|
IBA Lifesciences
5-5003-001 |
Strep-tag II | pCSG-IBA3 |
266.00 €
5µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15613868
|
IBA Lifesciences
5-5064-001 |
Strep-tag II, FLAG-tag | pCSG-IBA64 |
266.00 €
5µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15613878
|
IBA Lifesciences
5-5145-001 |
Twin-Strep-tag, His Tag (6x) | pCSG-IBA145 |
266.00 €
5µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15623858
|
IBA Lifesciences
5-5005-001 |
Strep-tag II | pCSG-IBA5 |
266.00 €
5µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15623878
|
IBA Lifesciences
5-5162-001 |
Twin-Strep-tag, FLAG-tag | pCSG-IBA162 |
266.00 €
5µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15633858
|
IBA Lifesciences
5-5023-001 |
Strep-tag II, GST Tag | pCSG-IBA23 |
266.00 €
5µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15633878
|
IBA Lifesciences
5-5164-001 |
Twin-Strep-tag, FLAG-tag | pCSG-IBA164 |
266.00 €
5µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15643868
|
IBA Lifesciences
5-5103-001 |
Twin-Strep-tag | pCSG-IBA103 |
266.00 €
5µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15643878
|
IBA Lifesciences
5-5167-001 |
FLAG-tag-Twin-Strep-tag | pCSG-IBA167 |
266.00 €
5µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15653878
|
IBA Lifesciences
5-5168-001 |
Twin-Strep-tag-FLAG-tag | pCSG-IBA168 |
266.00 €
5µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15663868
|
IBA Lifesciences
5-5105-001 |
Twin-Strep-tag | pCSG-IBA105 |
266.00 €
5µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15673858
|
IBA Lifesciences
5-5043-001 |
His Tag (6x), Strep-tag II | pCSG-IBA43 |
266.00 €
5µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15673868
|
IBA Lifesciences
5-5123-001 |
Twin-Strep-tag, GST Tag | pCSG-IBA123 |
266.00 €
5µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15683858
|
IBA Lifesciences
5-5045-001 |
His Tag (6x), Strep-tag II | pCSG-IBA45 |
266.00 €
5µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15693848
|
IBA Lifesciences
5-5000-001 |
Untagged | pCSG-IBAwt1 |
266.00 €
5µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15693858
|
IBA Lifesciences
5-5062-001 |
Strep-tag II, FLAG-tag | pCSG-IBA62 |
266.00 €
5µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15693868
|
IBA Lifesciences
5-5143-001 |
His Tag (6x), Twin-Strep-tag | pCSG-IBA143 |
266.00 €
5µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15653858
|
IBA Lifesciences
5-5033-001 |
His Tag (6x) | pCSG-IBA33 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15643858
|
IBA Lifesciences
5-5025-001 |
GST Tag | pCSG-IBA25 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15663858
|
IBA Lifesciences
5-5035-001 |
His Tag (6x) | pCSG-IBA35 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15623868
|
IBA Lifesciences
5-5065-001 |
FLAG-tag | pCSG-IBA65 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15603868
|
IBA Lifesciences
5-5063-001 |
FLAG-tag | pCSG-IBA63 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Designed for high-level episomal expression in mammalian hosts under G418 selection. IBA Lifesciences GmbH Mammalian Expression Vector with No Secretion is a large expression vector with universal features for transient expression as well as for generation of stable cell lines (NeoR).Specifications
Constitutive | |
CMV | |
Geneticin™ (G-418) | |
5 μg |
Ampicillin | |
Vector | |
Recommended: 2°C to 8°C for frequent usage, -20°C for long-term storage | |
Episomal expression (oriP, EBNA-1) |