missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences pCSG-IBA Mammalian Expression Vector with Secretion
GREENER_CHOICE

Product Code. 15603878 Shop All IBA Lifesciences Products
Click to view available options
Vector:
pCSG-IBA102
pCSG-IBA104
pCSG-IBA142
pCSG-IBA144
pCSG-IBAwt2
Protein Tag:
His Tag (6x), Twin-Strep-tag
Twin-Strep-tag
Twin-Strep-tag, His Tag (6x)
Untagged
Unit Size:
5µg
This item is not returnable. View return policy

Product Code. 15603878

missing translation for 'mfr': IBA Lifesciences 55144001

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Designed for high-level episomal expression in mammalian hosts under G418 selection. Mammalian Expression Vector with Secretion is a large expression vector with universal features for transient expression as well as for generation of stable cell lines (NeoR).

pCSG-IBA mammalian expression vectors
  • Designed for high-level episomal expression in mammalian hosts under G418 selection containing the following elements:
  • Human cytomegalovirus (CMV) immediate-early promoter for high-level expression in a wide range of mammalian cells
  • Episomal replication through Epstein Barr Virus replication origin (oriP) and nuclear antigen encoded by EBNA-1 in human, primate and canine cells and through SV40 replication origin in cells latently infected with SV40 or that express the SV40 large T antigen (e.g. HEK293T, COS-1, COS-7)
  • Stable cell lines can be selected by the neomycin resistance gene (NeoR).
  • In addition, the human cytomegalovirus (CMV) immediate-early promoter enables a high-level expression in a wide range of mammalian cells.

Please note that cloning into pCSG-IBA Acceptor Vectors compulsorily requires the restriction enzyme Esp3I since no other MCS for the integration of a gene of interest is available. Besides to the direct cloning of the gene of interest into pCSG-IBA vectors with Esp3I, another option via a so-called entry-vector is possible.

Specifications

Constitutive or Inducible System Constitutive
Marker Gene Ampicillin
Promoter CMV
Product Type Vector
Selection Agent (Eukaryotic) Geneticin™ (G-418)
Content And Storage Recommended: 2°C to 8°C for frequent usage, -20°C for long-term storage
Protein Tag Twin-Strep-tag, His Tag (6x)
Quantity 5 μg
Vector pCSG-IBA144
For Use With (Application) Episomal expression (oriP, EBNA-1); BM40 secretion signal
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.