missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mancozeb (Technical Grade), TRC
Especificaciones
Especificaciones
| Chemical Name or Material | Mancozeb |
| CAS | 8018-01-7 |
| Molecular Formula | C4 H6 Mn N2 S4 . C4 H6 N2 S4 Zn |
| Quantity | 1 g |
| Synonym | Manganese, [N-[2-[(dithiocarboxy)amino]ethyl]carbamodithioato(2-)-κS,κS']-, mixt. with [N-[2-[(dithiocarboxy)amino]ethyl]carbamodithioato(2-)-κS,κS']zinc (ACI), Carbamic acid, ethylenebis[dithio-, manganese zinc complex (8CI), Carbamodithioic acid, 1,2-ethanediylbis-, manganese complex (ZCI), Carbamodithioic acid, 1,2-ethanediylbis-, zinc complex (ZCI), Manganese, [[1,2-ethanediylbis[carbamodithioato]](2-)]-, mixt. with [[1,2-ethanediylbis[carbamodithioato]](2-)]zinc (ZCI), Manganese, [[2-[(dithiocarboxy)amino]ethyl]carbamodithioato(2-)-κS,κS']-, mixt. with [[2-[(dithiocarboxy)amino]ethyl]carbamodithioato(2-)-κS,κS']zinc (9CI), Zinc, [[1,2-ethanediylbis[carbamodithioato]](2-)]-, mixt. contg. (ZCI), Zinc, [[2-[(dithiocarboxy)amino]ethyl]carbamodithioato(2-)-κS,κS']-, mixt. contg. (9CI), Agrox 16D, Agrozeb 80WP, Banazeb 60SC, Carmazine, Contizeb, Daisenmengxin, Dhanuka M 45, Diesen M, Dithane, Dithane 60SC, Dithane 945, Dithane DG, Dithane DG NT, Dithane F, Dithane F 45, Dithane M 45, Dithane M45, Dithane M80, Dithane Neo Tec 75WG, Dithane NT, Dithane OS, Dithane Rainshield Neo Tec, Dithane S 60, Dithane SPC, Dithane ultra, F 2966, Fore, Green-daisen M, Greenzeb 80WP, Hi-shield, Indofil 80WP, Indofil M 45, Indofil M45, Jimanaisen, Jimandaisen, Karamate, Karamate N, Kascade, Liro manzeb, Mancomix, Mancosan, Mancozan, Mancozeb, Mancozebe Sipcam, Mancozi, Maneb-zinc, Mankogal, Manoseb, Manzate 200, Manzeb, Manzidan, Marzin, Milcozebe, Mount 45, Mycotrin, Nemispor, Novozir MN, Novozir MN 80, Othane M 45, Pencozeb, Penncozeb, Pennfluid, Sakozeb Blue, Sancozeb, Sandozeb Pepite, Suncozeb, Tanzeo M 45, Thane M 45, Trimanoc, Tritogol MZ, Vondozeb, Ziman-Dithane, Zimanat, Zimaneb, Zinc manganese ethylenebisdithiocarbamate |
| InChI Formula | InChI=1S/2C4H8N2S4.Mn.Zn/c2*7-3(8)5-1-2-6-4(9)10;;/h2*1-2H2,(H2,5,7,8)(H2,6,9,10);;/q;;2*+2/p-4 |
| SMILES | S=C1NCCNC(=S)S[Zn]S1.S=C2NCCNC(=S)S[Mn]S2 |
| IUPAC Name | 1,3-dithia-5,8-diaza-2λ^{2}-manganacyclononane-4,9-dithione;1,3-dithia-5,8-diaza-2λ^{2}-zincacyclononane-4,9-dithione |
| Molecular Weight (g/mol) | 541.07 |
| Formula Weight | 538.75 |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?