missing translation for 'onlineSavingsMsg'
Learn More

Melford Manganese Chloride 1M Solution

Product Code. 30018457
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018457 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30018457 Supplier Melford Supplier No. M2010050.0
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Manganese Chloride is used for dC and dG tailing of DNA.

Manganese Chloride is used for dC and dG tailing of DNA.

Specifications

Chemical Name or Material Manganese Chloride 1 M Solution
CAS 7732-18-5, 7773-01-5
Quantity 50 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.