missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) 2,4-pentanedionate

Product Code. 11453038
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11453038 500 g 500g
11433038 25 g 25g
11443038 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11453038 Supplier Thermo Scientific Alfa Aesar Supplier No. A18762.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14024-58-9 | C10H14MnO4 | 253.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14024-58-9
Molecular Formula C10H14MnO4
Molecular Weight (g/mol) 253.16
MDL Number MFCD00000022 MFCD09998212
InChI Key ZQZQURFYFJBOCE-FDGPNNRMSA-L
Synonym bis 4-hydroxypent-3-en-2-one dihydrate manganese
PubChem CID 54669727
IUPAC Name (Z)-4-hydroxypent-3-en-2-one;manganese;dihydrate
SMILES [Mn++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 260°C to 262°C (decomposition)
Density 1.6
Quantity 500 g
Formula Weight 253.16
Chemical Name or Material Manganese(II) 2,4-pentanedionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.