missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) acetate tetrahydrate, 99+%, for analysis

Product Code. 10700911 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
2.5kg
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10700911 25 g Plastic bottle 25g
10082740 500 g Plastic bottle 500g
10723475 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10700911 Supplier Thermo Scientific Acros Supplier No. 205660250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 6156-78-1 | C4H14MnO8 | 245.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6156-78-1
Molecular Formula C4H14MnO8
Molecular Weight (g/mol) 245.09
MDL Number MFCD00062552
InChI Key CESXSDZNZGSWSP-UHFFFAOYSA-L
Synonym manganese ii acetate tetrahydrate, manganese acetate tetrahydrate, manganous acetate tetrahydrate, unii-9to51d176n, manganese diacetate, tetrahydrate, manganese 2+ diacetate tetrahydrate, acetic acid, manganese 2+ salt, tetrahydrate, manganese ii acetatetetrahydrate, acmc-20akkp
PubChem CID 93021
SMILES O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point >300.0°C
Color Pink
pH 6 to 7 (5% soln.)
Physical Form Crystals
Quantity 25 g
Assay Percent Range 99% min. (Complexometry)
Linear Formula (CH3CO2)2Mn·4H2O
Flash Point >130°C
Merck Index 14, 5724
Solubility Information Solubility in water: very soluble. Other solubilities: soluble in alcohol
IUPAC Name manganese(2+) diacetate tetrahydrate
Formula Weight 245.09
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Density 1.58
Chemical Name or Material Manganese(II) acetate tetrahydrate, 99+%
Show More Show Less
Product Identifier
  • Manganese(II) acetate tetrahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.