missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) acetate tetrahydrate, Mn 22% (typical)

Product Code. 11391719
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
Product Code. Quantity unitSize
11391719 500 g 500g
11301729 100 g 100g
11311729 2 kg 2kg
3 options
This item is not returnable. View return policy

Product Code. 11391719

Brand: Thermo Scientific Alfa Aesar 012351.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6156-78-1 | C4H14MnO8 | 245.09 g/mol

It is used as a mordant in dyeing, drier for paints and varnishes, an oxidation catalyst, and in the manufacture of bister. It is also used in the manufacture of fertilizer. It is used for the production of high purity manganese dioxide, manganese oxide hydrate, oxide-based catalysts, and catalysts for oxidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Mordant in dyeing, in the manufacture of bister, as a drier for paints and varnishes, and as an oxidation catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6156-78-1
Molecular Formula C4H14MnO8
Molecular Weight (g/mol) 245.09
MDL Number MFCD00062552
InChI Key CESXSDZNZGSWSP-UHFFFAOYSA-L
Synonym manganese ii acetate tetrahydrate, manganese acetate tetrahydrate, manganous acetate tetrahydrate, unii-9to51d176n, manganese diacetate, tetrahydrate, manganese 2+ diacetate tetrahydrate, acetic acid, manganese 2+ salt, tetrahydrate, manganese ii acetatetetrahydrate, acmc-20akkp
PubChem CID 93021
IUPAC Name manganese(2+);diacetate;tetrahydrate
SMILES O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 80°C
Color Pink
Density 1.589 g/mL
Odor Pungent
Linear Formula Mn(OOCCH3)2·4H2O
Quantity 500 g
Merck Index 14,5724
Solubility Information Soluble in water,methanol,ethanol and acetic acid.
Formula Weight 245.09 (173.03 Anhydrous)
Concentration or Composition (by Analyte or Components) Mn 22% (typical)
Physical Form Crystalline
Chemical Name or Material Manganese(II) acetate tetrahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.