missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) bromide, anhydrous, 95%

Product Code. 11433887
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. Quantity unitSize
11433887 100 g 100g
11423887 25 g 25g
2 options
This item is not returnable. View return policy

Product Code. 11433887

Brand: Thermo Scientific Alfa Aesar A18321.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13446-03-2 | MnBr2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-03-2
Molecular Formula MnBr2
MDL Number MFCD00011113

Specifications

Melting Point 698°C
Quantity 100 g
Merck Index 14,5725
Formula Weight 214.76
Percent Purity 95%
Odor Odorless
Sensitivity Hygroscopic
Density 4.38
Chemical Name or Material Manganese(II) bromide, anhydrous

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.