missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) bromide, anhydrous, 99%, H{2}O <0.3%

Product Code. 11382988
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382988 10 g 10g
11372988 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382988 Supplier Thermo Scientific Alfa Aesar Supplier No. 032736.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13446-03-2

It can be used in place of palladium in the Stille reaction, which couples two carbon atoms using an organotin compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-03-2
MDL Number MFCD00011113

Specifications

Melting Point 698°C
Physical Form Powder
Quantity 10 g
Merck Index 14,5725
Solubility Information Soluble in water. (1273g/L)
Formula Weight 214.76
Percent Purity 99%
Concentration or Composition (by Analyte or Components) H2O <0.3%
Odor Odorless
Sensitivity Hygroscopic
Density 4.38 g/mL
Chemical Name or Material Manganese(II) bromide, Anhydrous
Show More Show Less
compliance-icons
Product Identifier
  • Manganese(II) bromide, anhydrous
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Manganese bromide (MnBr2)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.