missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) bromide tetrahydrate, 98%

Product Code. 10134120 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
Packaging:
Glass bottle
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10134120 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10134120 Supplier Thermo Scientific Acros Supplier No. 197130500
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10031-20-6 | Br2H8MnO4 | 286.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 10031-20-6
Color Pink
Physical Form Crystalline Powder and/or Crystals
Quantity 50 g
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula Br2H8MnO4
Linear Formula MnBr2·4H2O
MDL Number MFCD00149791
Merck Index 14, 5725
Synonym manganese bromide tetrahydrate, unii-8308v6rbj5, manganese ii bromide hexahydrate, manganous bromide tetrahydrate, manganese dibromide tetrahydrate, manganese bromide mnbr2 , tetrahydrate, manganese 2+ ion tetrahydrate dibromide
Solubility Information Solubility in water: soluble
InChI Key HHDPJRSXPOGIOP-UHFFFAOYSA-L
SMILES O.O.O.O.[Mn++].[Br-].[Br-]
IUPAC Name manganese(2+) tetrahydrate dibromide
Molecular Weight (g/mol) 286.81
Formula Weight 286.82
Percent Purity 98%
Chemical Name or Material Manganese(II) bromide tetrahydrate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Manganese(II) bromide tetrahydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.