missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) carbonate, Puratronic™, 99.985% (metals basis)

Product Code. 11355057
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11355057 100 g 100g
11345057 5 g 5g
11335057 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11355057 Supplier Thermo Scientific Alfa Aesar Supplier No. 040756.22
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 598-62-9 | CMnO3 | 114.946 g/mol

Manganese(II) carbonate is used in the production of manganese dioxide for dry-cell batteries and for ferrites, as an additive to plant fertilizers to cure manganese deficient crops. It is also used in health foods, in ceramics as a glaze colorant and flux, and in concrete stains, and as a hematinic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-62-9
Molecular Formula CMnO3
Molecular Weight (g/mol) 114.946
MDL Number MFCD00011116
InChI Key XMWCXZJXESXBBY-UHFFFAOYSA-L
Synonym manganese carbonate, manganese ii carbonate, manganous carbonate, manganese 2+ carbonate, natural rhodochrosite, manganese carbonate 1:1, rhodochrosite, manganese carbonate mnco3, unii-9zv57512zm, ccris 3660
PubChem CID 11726
IUPAC Name manganese(2+);carbonate
SMILES C(=O)([O-])[O-].[Mn+2]

Specifications

Melting Point 350°C (decomposition)
Color Brown to Violet
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Soluble in water(0.065g/L),dilute inorganic acids. Insoluble alcohol.
Formula Weight 114.95
Odor Odorless
Appearance Light brown to violet powder
Density 3.125
Chemical Name or Material Manganese(II) carbonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.