missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) chloride, 97%

Product Code. 10405755 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Plastic Drum
Unit Size:
1kg
2.5kg
250g
25g
Product Code. Quantity Packaging unitSize
10405755 250 g Glass bottle 250g
10093320 25 g Glass bottle 25g
10698191 1 kg Glass bottle 1kg
10687734 2.5 kg Plastic Drum 2.5kg
4 options
This item is not returnable. View return policy

Product Code. 10405755

Brand: Thermo Scientific Acros 271412500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7773-01-5 | Cl2Mn | 125.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7773-01-5
Molecular Formula Cl2Mn
Molecular Weight (g/mol) 125.84
MDL Number MFCD00011114
InChI Key GLFNIEUTAYBVOC-UHFFFAOYSA-L
Synonym manganese chloride, manganese ii chloride, manganese 2+ ion dichloride, manganesechloride, acmc-20akkt, manganous chloride,anhydrous, ksc171o6h, manganous chloride, anhydr, manganese ii chloride anhydrous crystalline
PubChem CID 10313134
ChEBI CHEBI:63041
SMILES [Cl-].[Cl-].[Mn++]

Specifications

Boiling Point 1190.0°C
Melting Point 650.0°C
Color Pink
Physical Form Flakes
Quantity 250 g
Assay Percent Range 96% min. (Complexometry)
Linear Formula MnCl2
Fieser 07,222
Merck Index 15, 5793
Solubility Information Solubility in water: 723g/L (25°C). Other solubilities: soluble in alcohol,insoluble in ether
IUPAC Name manganese(2+);dichloride
Formula Weight 125.84
Percent Purity 97%
Packaging Glass bottle
Chemical Name or Material Manganese(II) chloride
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Manganese(II) chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H373-May cause damage to organs.
Precautionary Statement
  • P270-Do not eat, drink or smoke when using this product.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.