missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(II) chloride tetrahydrate, 99+%, for analysis
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
| Melting Point | 58.0°C |
| Color | Pink |
| Physical Form | Fine Crystalline Powder |
| Quantity | 2.5 kg |
| Assay Percent Range | 99% min. (Complexometry) |
| Linear Formula | MnCl2·4H2O |
| Fieser | 07,222; 09,288 |
| Merck Index | 15,5793 |
| Solubility Information | Solubility in water: 1980g/L (20°C). Other solubilities: soluble in alcohol, insoluble in ether |
| Formula Weight | 197.91 |
| Mehr anzeigen |
Sicherheit und Handhabung
Produktidentifikator
- Manganese(II) chloride tetrahydrate
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity after repeated exposure Category 2
- H318-Causes serious eye damage.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur