missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) Chloride Tetrahydrate, Certified AR for Analysis, Fisher Chemical™

Product Code. 10181620 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10181620 500 g 500g
10376090 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10181620 Supplier Fisher Chemical Supplier No. M/1850/53
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 13446-34-9
Molecular Formula Cl2H8MnO4
Molecular Weight (g/mol) 197.90
MDL Number MFCD00149792
InChI Key CNFDGXZLMLFIJV-UHFFFAOYSA-L
IUPAC Name manganese(2+) tetrahydrate dichloride
SMILES O.O.O.O.[Cl-].[Cl-].[Mn++]

Specifications

Melting Point 58°C
Color Red
pH 4 to 6
Physical Form Powder/Solid
Quantity 500 g
Formula Weight 197.91g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Manganese(II) chloride tetrahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.